2,5-Dibromo-4-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

65160-1G 65160-25G 65160-500MG 65160-5G
76905-692EA 203.7 USD
76905-692 76905-694 76905-696 76905-698
2,5-Dibromo-4-fluorobenzoic acid ≥97%
2,5-Dibromo-4-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃Br₂FO₂
Storage Temperature: Ambient
CAS Number: 187326-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR