2-Bromo-4-isopropoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

64978-1G 64978-25G 64978-500MG 64978-5G
76905-676EA 292.81 USD
76905-676 76905-678 76905-680 76905-682
2-Bromo-4-isopropoxybenzaldehyde ≥97%
2-Bromo-4-isopropoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
MW: 243.0971 g/mol
Storage Temperature: Refrigerator
CAS Number: 1289064-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR