2-Chloro-3-iodophenol ≥97% research grade

Supplier: AOBChem USA

Warning

64781-1G 64781-25G 64781-5G
76905-664EA 62.6 USD
76905-664 76905-666 76905-668
2-Chloro-3-iodophenol ≥97% research grade
2-Chloro-3-iodophenol
Formula: C₆H₄ClIO
Storage Temperature: Refrigerator
MDL Number: MFCD08166415
CAS Number: 666727-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR