2-Fluoro-3-Methoxy-1-(Methylsulfonyl)-4-(Trifluoromethyl)Benzene ≥97%
Supplier: AOBChem USA
|
Warning
|
63807-1G
63807-25G
63807-500MG
63807-5G
76905-648EA
240.83
USD
76905-648
76905-650
76905-652
76905-654
2-Fluoro-3-Methoxy-1-(Methylsulfonyl)-4-(Trifluoromethyl)Benzene ≥97%
2-Fluoro-3-Methoxy-1-(Methylsulfonyl)-4-(Trifluoromethyl)Benzene
Precaución: for Research Use Only
Formula:
C₉H₈F₄O₃S MW: 272.2166 g/mol Storage Temperature: Ambient |
CAS Number:
2807469-49-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...