2,6-Difluoro-3,5-dimethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

63704-1G 63704-25G 63704-500MG 63704-5G
76905-632EA 266.29 USD
76905-632 76905-634 76905-636 76905-638
2,6-Difluoro-3,5-dimethoxybenzoic acid ≥97%
2,6-Difluoro-3,5-dimethoxybenzoic acid
Formula: C₉H₈F₂O₄
Storage Temperature: Ambient
MDL Number: MFCD24448610
CAS Number: 651734-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR