2-Methoxy-6-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

63302-1G 63302-25G 63302-5G
76905-620EA 64.72 USD
76905-620 76905-622 76905-624
2-Methoxy-6-methylbenzaldehyde ≥97%
2-Methoxy-6-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
MW: 150.174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08235006
CAS Number: 54884-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR