2-Ethoxy-6-methylaniline ≥97%

Supplier: AOBChem USA

Warning

63232-1G 63232-25G 63232-500MG 63232-5G
76905-604EA 177.18 USD
76905-604 76905-606 76905-608 76905-610
2-Ethoxy-6-methylaniline ≥97%
2-Ethoxy-6-methylaniline
Formula: C₉H₁₃NO
MW: 151.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12170060
CAS Number: 53982-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR