2-Iodo-1-methoxy-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

63211-1G 63211-25G 63211-500MG 63211-5G
76905-588EA 423.3 USD
76905-588 76905-590 76905-592 76905-594
2-Iodo-1-methoxy-3-methylbenzene ≥97%
2-Iodo-1-methoxy-3-methylbenzene
Formula: C₈H₉IO
MW: 248.0609 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18397767
CAS Number: 35387-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR