2,6-Dibromo-4-chloro-3-methylaniline ≥97%

Supplier: AOBChem USA

Warning

63188-25G 63188-5G
76905-576EA 552.73 USD
76905-576 76905-578
2,6-Dibromo-4-chloro-3-methylaniline ≥97%
2,6-Dibromo-4-chloro-3-methylaniline

Precaución: for Research Use Only

Formula: C₇H₆Br₂ClN
Storage Temperature: Ambient
CAS Number: 861559-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR