2-Bromo-4-fluoro-6-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

62704-25G 62704-5G
76905-566EA 524.09 USD
76905-566 76905-568
2-Bromo-4-fluoro-6-methoxyphenol ≥97%
2-Bromo-4-fluoro-6-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₆BrFO₂
Storage Temperature: Ambient
CAS Number: 1782378-34-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR