2-Fluoro-5-methoxy-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

62464-1G 62464-25G 62464-500MG 62464-5G
76905-542EA 203.7 USD
76905-542 76905-544 76905-546 76905-548
2-Fluoro-5-methoxy-3-methylbenzoic acid ≥97%
2-Fluoro-5-methoxy-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MW: 184.1643 g/mol
Storage Temperature: Ambient
CAS Number: 447463-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR