2,6-Dibromo-3-fluoro-4-methylaniline ≥97%

Supplier: AOBChem USA

Warning

62229-1G 62229-25G 62229-5G
76905-524EA 77.45 USD
76905-524 76905-526 76905-528
2,6-Dibromo-3-fluoro-4-methylaniline ≥97%
2,6-Dibromo-3-fluoro-4-methylaniline

Precaución: for Research Use Only

Formula: C₇H₆Br₂FN
Storage Temperature: Ambient
MDL Number: MFCD09800769
CAS Number: 1000576-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR