2,6-Dichloro-3-fluoro-4-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

62033-1G 62033-25G 62033-500MG 62033-5G
76905-498EA 203.7 USD
76905-498 76905-500 76905-502 76905-504
2,6-Dichloro-3-fluoro-4-iodopyridine ≥97%
2,6-Dichloro-3-fluoro-4-iodopyridine

Precaución: for Research Use Only

Formula: C₅HCl₂FIN
Storage Temperature: Ambient
CAS Number: 2756590-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR