2-Bromo-6-chloro-3-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

61367-1G 61367-25G 61367-500MG 61367-5G
76905-466EA 346.92 USD
76905-466 76905-468 76905-470 76905-472
2-Bromo-6-chloro-3-methoxypyridine ≥97%
2-Bromo-6-chloro-3-methoxypyridine
Formula: C₆H₅BrClNO
Storage Temperature: Ambient
MDL Number: MFCD18255399
CAS Number: 1256819-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR