2-Bromo-4-chloro-1,3,5-trifluorobenzene ≥97%

Supplier: AOBChem USA

Warning

61117-25G 61117-5G
76905-454EA 516.66 USD
76905-454 76905-456
2-Bromo-4-chloro-1,3,5-trifluorobenzene ≥97%
2-Bromo-4-chloro-1,3,5-trifluorobenzene

Precaución: for Research Use Only

Formula: C₆HBrClF₃
MW: 245.4243 g/mol
Storage Temperature: Ambient
CAS Number: 292621-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR