2-Chloro-4-isopropoxy-5-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

60588-1G 60588-25G 60588-5G 60588-500MG
76905-418EA 84.88 USD
76905-418 76905-420 76905-422 77647-804
2-Chloro-4-isopropoxy-5-methoxybenzoic acid ≥97%
2-Chloro-4-isopropoxy-5-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClO₄
MW: 244.672 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05996113
CAS Number: 713104-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR