2-Chloro-4-(hydroxymethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

60245-1G 60245-25G 60245-500MG 60245-5G
76905-392EA 319.34 USD
76905-392 76905-394 76905-396 76905-398
2-Chloro-4-(hydroxymethyl)phenol ≥97%
2-Chloro-4-(hydroxymethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₇ClO₂
Storage Temperature: Refrigerator
CAS Number: 105960-29-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR