2-Ethoxy-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

60126-25G 60126-5G
76905-380EA 310.85 USD
76905-380 76905-382
2-Ethoxy-3-methoxybenzaldehyde ≥97%
2-Ethoxy-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Storage Temperature: Ambient
CAS Number: 66799-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR