3,4-Dimethoxysalicylic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Hydroxy-3,4-dimethoxybenzoic acid

60092-1G 60092-25G 60092-500MG 60092-5G
76905-364EA 124.13 USD
76905-364 76905-366 76905-368 76905-370
3,4-Dimethoxysalicylic acid ≥97%
3,4-Dimethoxysalicylic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₅
MW: 198.18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00016498
CAS Number: 5653-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR