2-Methoxypyrimidin-5-ol ≥97%

Supplier: AOBChem USA

Warning

59445-1G 59445-25G 59445-500MG 59445-5G
76905-348EA 135.8 USD
76905-348 76905-350 76905-352 76905-354
2-Methoxypyrimidin-5-ol ≥97%
2-Methoxypyrimidin-5-ol

Precaución: for Research Use Only

Formula: C₅H₆N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD10696950
CAS Number: 91233-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR