2-Chloro-4-fluoro-3-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

58990-1G 58990-25G 58990-500MG 58990-5G
76905-332EA 259.93 USD
76905-332 76905-334 76905-336 76905-338
2-Chloro-4-fluoro-3-formylbenzoic acid ≥97%
2-Chloro-4-fluoro-3-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄ClFO₃
MW: 202.567 g/mol
Storage Temperature: Ambient
CAS Number: 2385557-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR