2-Ethoxy-5-fluoro-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

58344-1G 58344-500MG 58344-5G
76905-294EA 654.58 USD
76905-294 76905-296 76905-298
2-Ethoxy-5-fluoro-3-methylbenzaldehyde ≥97%
2-Ethoxy-5-fluoro-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₂
Storage Temperature: Ambient
CAS Number: 883537-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR