1,4-Dibromo-3,5-difluorobenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,5-Dibromo-1,3-difluorobenzene

57654-25G 57654-5G
76905-282EA 194.15 USD
76905-282 76905-284
1,4-Dibromo-3,5-difluorobenzene ≥97%
1,4-Dibromo-3,5-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₂F₂
MW: 271.8849 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07783665
CAS Number: 128259-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR