2-Chloro-3-(trifluoromethyl)benzamide ≥95%

Supplier: AOBChem USA

Warning

56946-25G 56946-5G
76905-258EA 418 USD
76905-258 76905-260
2-Chloro-3-(trifluoromethyl)benzamide ≥95%
2-Chloro-3-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₈H₅ClF₃NO
Storage Temperature: Ambient
MDL Number: MFCD04115831
CAS Number: 39959-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR