2,6-Difluoro-4-iodobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

56933-1G 56933-25G 56933-500MG 56933-5G
76905-242EA 257.8 USD
76905-242 76905-244 76905-246 76905-248
2,6-Difluoro-4-iodobenzoic acid ≥95%
2,6-Difluoro-4-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃F₂IO₂
MW: 283.999 g/mol
Storage Temperature: Ambient
CAS Number: 1160575-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR