2-Chloro-5-iodo-1,3-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

56381-1G 56381-25G 56381-5G
76905-212EA 62.6 USD
76905-212 76905-214 76905-216
2-Chloro-5-iodo-1,3-dimethylbenzene ≥95%
2-Chloro-5-iodo-1,3-dimethylbenzene

Precaución: for Research Use Only

Formula: C₈H₈ClI
MW: 266.507 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530519
CAS Number: 294662-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR