2-Bromo-5-iodobenzamide ≥97%

Supplier: AOBChem USA

Warning

56229-1G 56229-25G 56229-500MG 56229-5G
76905-196EA 144.29 USD
76905-196 76905-198 76905-200 76905-202
2-Bromo-5-iodobenzamide ≥97%
2-Bromo-5-iodobenzamide

Precaución: for Research Use Only

Formula: C₇H₅BrINO
Storage Temperature: Ambient
CAS Number: 865178-41-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR