2-Aminopyrimidin-5-ol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Amino-5-hydroxypyrimidine

55598-1G 55598-25G 55598-500MG 55598-5G
76905-162EA 173.99 USD
76905-162 76905-164 76905-166 76905-168
2-Aminopyrimidin-5-ol ≥97%
2-Aminopyrimidin-5-ol
Formula: C₄H₅N₃O
MW: 111.1 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08062411
CAS Number: 143489-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR