2-Bromo-6-chloro-pyridine-3-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

55187-1G 55187-25G 55187-500MG 55187-5G
76905-146EA 296 USD
76905-146 76905-148 76905-150 76905-152
2-Bromo-6-chloro-pyridine-3-carbaldehyde ≥97%
2-Bromo-6-chloro-pyridine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₆H₃BrClNO
Storage Temperature: Ambient
MDL Number: MFCD11844804
CAS Number: 1060815-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR