2-Bromo-6-chloro-3-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

55180-1G 55180-25G 55180-500MG 55180-5G
76905-130EA 292.81 USD
76905-130 76905-132 76905-134 76905-136
2-Bromo-6-chloro-3-iodopyridine ≥97%
2-Bromo-6-chloro-3-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClIN
Storage Temperature: Refrigerator
MDL Number: MFCD18257120
CAS Number: 1211580-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR