2-Fluoro-5-iodobenzamide ≥97%

Supplier: AOBChem USA

Warning

55028-1G 55028-25G 55028-500MG 55028-5G
76905-114EA 274.78 USD
76905-114 76905-116 76905-118 76905-120
2-Fluoro-5-iodobenzamide ≥97%
2-Fluoro-5-iodobenzamide

Precaución: for Research Use Only

Formula: C₇H₅FINO
MW: 265.024 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18396918
CAS Number: 866683-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR