2,5-Dichloro-4-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

54785-1G 54785-25G 54785-500MG 54785-5G
76905-088EA 198.39 USD
76905-088 76905-090 76905-092 76905-094
2,5-Dichloro-4-methoxybenzoic acid ≥97%
2,5-Dichloro-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00214270
CAS Number: 2500-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR