6-Methoxy-o-cresol ≥97%

Supplier: AOBChem USA

Warning

54724-25G 54724-5G
76905-062EA 212.18 USD
76905-062 76905-064
6-Methoxy-o-cresol ≥97%
6-Methoxy-o-cresol

Precaución: for Research Use Only

Formula: C₈H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00043771
CAS Number: 2896-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR