2-Bromo-5-chloro-1-methyl-3-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

54198-1G 54198-25G 54198-5G
76905-028EA 68.96 USD
76905-028 76905-030 76905-032
2-Bromo-5-chloro-1-methyl-3-nitrobenzene ≥97%
2-Bromo-5-chloro-1-methyl-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅BrClNO₂
Storage Temperature: Ambient
MDL Number: MFCD28734432
CAS Number: 631910-13-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR