2-Chloro-5-methylisonicotinic acid ≥97%

Supplier: AOBChem USA

Warning

54132-1G 54132-500MG 54132-5G
76905-018EA 643.97 USD
76905-018 76905-020 76905-022
2-Chloro-5-methylisonicotinic acid ≥97%
2-Chloro-5-methylisonicotinic acid
Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13188984
CAS Number: 951030-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR