2-Bromo-6-chloro-3-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

53823-25G 53823-5G
76904-996EA 453.01 USD
76904-996 76904-998
2-Bromo-6-chloro-3-methoxyphenol ≥97%
2-Bromo-6-chloro-3-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₆BrClO₂
Storage Temperature: Ambient
MDL Number: MFCD34564243
CAS Number: 2092775-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR