2-Bromo-4-(1,1-difluoroethyl)-1-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

53462-1G 53462-25G 53462-5G
76904-956EA 75.33 USD
76904-956 76904-958 76904-960
2-Bromo-4-(1,1-difluoroethyl)-1-fluorobenzene ≥97%
2-Bromo-4-(1,1-difluoroethyl)-1-fluorobenzene
Formula: C₈H₆BrF₃
Storage Temperature: Ambient
MDL Number: MFCD11845795
CAS Number: 1138444-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR