2-Chloro-4-iodo-5-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

53413-1G 53413-25G 53413-500MG 53413-5G
76904-942EA 153.84 USD
76904-942 76904-944 76904-946 76904-948
2-Chloro-4-iodo-5-methylpyridine ≥97%
2-Chloro-4-iodo-5-methylpyridine
Formula: C₆H₅ClIN
Storage Temperature: Refrigerator
MDL Number: MFCD16610529
CAS Number: 1197957-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR