2-Bromo-4-chloro-1,3-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

53191-1G 53191-25G 53191-500MG 53191-5G
76904-902EA 332.07 USD
76904-902 76904-904 76904-906 76904-908
2-Bromo-4-chloro-1,3-dimethylbenzene ≥95%
2-Bromo-4-chloro-1,3-dimethylbenzene
Formula: C₈H₈BrCl
MW: 219.50 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12922559
CAS Number: 1208077-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR