2-Fluoro-4-iodo-1-methoxybenzene ≥97% research grade

Supplier: AOBChem USA

Warning

53040-1G 53040-25G 53040-5G
76904-874EA 62.6 USD
76904-874 76904-876 76904-878
2-Fluoro-4-iodo-1-methoxybenzene ≥97% research grade
2-Fluoro-4-iodo-1-methoxybenzene
Formula: C₇H₆FIO
Storage Temperature: Refrigerator
MDL Number: MFCD00691526
CAS Number: 3824-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR