2,6-Dichloro-4-methoxybenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
52836-1G
52836-25G
52836-500MG
52836-5G
76904-858EA
411.63
USD
76904-858
76904-860
76904-862
76904-864
2,6-Dichloro-4-methoxybenzoic acid ≥97%
2,6-Dichloro-4-methoxybenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₆Cl₂O₃ Storage Temperature: Ambient |
CAS Number:
94278-69-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...