2-Bromo-4,5-dichlorobenzoic acid ≥97%

Supplier: AOBChem USA

52320-1G 52320-500MG 52320-5G
76904-790EA 559.1 USD
76904-790 76904-792 76904-794
2-Bromo-4,5-dichlorobenzoic acid ≥97%
2-Bromo-4,5-dichlorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrCl₂O₂
MDL Number: MFCD28737829
CAS Number: 93361-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR