2-Bromo-3-fluoro-6-methylphenol ≥97%

Supplier: AOBChem USA

Warning

52291-1G 52291-25G 52291-500MG 52291-5G
76904-774EA 272.66 USD
76904-774 76904-776 76904-778 76904-780
2-Bromo-3-fluoro-6-methylphenol ≥97%
2-Bromo-3-fluoro-6-methylphenol

Precaución: for Research Use Only

Formula: C₇H₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD22414712
CAS Number: 1522001-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR