2-Bromo-2,2-difluoro-1-morpholinoethanone ≥97%

Supplier: AOBChem USA

Warning

52252-1G 52252-25G 52252-500MG 52252-5G
76904-748EA 123.07 USD
76904-748 76904-750 76904-752 76904-754
2-Bromo-2,2-difluoro-1-morpholinoethanone ≥97%
2-Bromo-2,2-difluoro-1-morpholinoethanone

Precaución: for Research Use Only

Formula: C₆H₈BrF₂NO₂
MW: 244.034 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22548902
CAS Number: 149229-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR