2-Bromo-2,2-difluoro-N-(4-methoxyphenyl)acetamide ≥97%

Supplier: AOBChem USA

Warning

52248-1G 52248-25G 52248-500MG 52248-5G
76904-732EA 233.4 USD
76904-732 76904-734 76904-736 76904-738
2-Bromo-2,2-difluoro-N-(4-methoxyphenyl)acetamide ≥97%
2-Bromo-2,2-difluoro-N-(4-methoxyphenyl)acetamide

Precaución: for Research Use Only

Formula: C₉H₈BrF₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD33404028
CAS Number: 1812869-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR