2-Bromo-2,2-difluoro-N-(p-tolyl)acetamide ≥97%

Supplier: AOBChem USA

Warning

52237-1G 52237-25G 52237-500MG 52237-5G
76904-716EA 350.1 USD
76904-716 76904-718 76904-720 76904-722
2-Bromo-2,2-difluoro-N-(p-tolyl)acetamide ≥97%
2-Bromo-2,2-difluoro-N-(p-tolyl)acetamide

Precaución: for Research Use Only

Formula: C₉H₈BrF₂NO
MW: 264.067 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902996
CAS Number: 1949728-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR