2-Chloro-3-fluoro-4-iodo-1-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

51536-1G 51536-25G 51536-500MG 51536-5G
76904-678EA 263.11 USD
76904-678 76904-680 76904-682 76904-684
2-Chloro-3-fluoro-4-iodo-1-methoxybenzene ≥97%
2-Chloro-3-fluoro-4-iodo-1-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅ClFIO
MW: 286.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902954
CAS Number: 2385211-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR