2-Methoxy-3-methylnaphthalene ≥95%

Supplier: AOBChem USA

Warning

51455-1G 51455-25G 51455-5G
76904-666EA 88.06 USD
76904-666 76904-668 76904-670
2-Methoxy-3-methylnaphthalene ≥95%
2-Methoxy-3-methylnaphthalene

Precaución: for Research Use Only

Formula: C₁₂H₁₂O
Storage Temperature: Ambient
MDL Number: MFCD17012351
CAS Number: 61873-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR