2-Fluoro-3-iodo-5-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

51215-1G 51215-500MG 51215-5G
76904-638EA 736.27 USD
76904-638 76904-640 76904-642
2-Fluoro-3-iodo-5-(methylthio)pyridine ≥97%
2-Fluoro-3-iodo-5-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₆H₅FINS
MW: 269.078 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28796419
CAS Number: 1806345-87-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR