2-Fluoro-3-methyl-5-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

50760-1G 50760-500MG 50760-5G
76904-610EA 525.15 USD
76904-610 76904-612 76904-614
2-Fluoro-3-methyl-5-(methylthio)benzoic acid ≥97%
2-Fluoro-3-methyl-5-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₂S
Storage Temperature: Ambient
MDL Number: MFCD22490197
CAS Number: 1822662-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR